芥子酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 白芥酸, 3,5-二甲氧基-4-羟基肉桂酸, 4-羟基-3,5-二甲氧基肉桂酸

D1765-25G D1765-5G
TCIAD1765-25GEA 0 CNY
TCIAD1765-25G TCIAD1765-5G
芥子酸 ≥98.0% (by GC, titration analysis)
芥子酸
分子式: C₁₁H₁₂O₅
分子量: 224.21 g/mol
沸点: 378
MDL编号: MFCD00004401
CAS号: 530-59-6
EINECS: 208-487-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c39cd23bc059909474aac093fb93020b5e1fa35c&vid=c39cd23bc059909474aac093fb93020b5e1fa35c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR