吲哚-2-羧酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
Synonyms:
2-羧基吲哚
I0332-25G
I0332-5G
TCIAI0332-25GEA
0
CNY
TCIAI0332-25G
TCIAI0332-5G
吲哚-2-羧酸 ≥98.0% (by GC, titration analysis)
吲哚-2-羧酸
分子式:
C₉H₇NO₂ 分子量: 161.16 g/mol |
MDL编号:
MFCD00005611 CAS号: 1477-50-5 EINECS: 216-030-4 |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...