水杨酸乙酯 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-羟基苯甲酸乙酯

S0011-25ML S0011-500ML
TCIAS0011-25MLEA 0 CNY
TCIAS0011-25ML TCIAS0011-500ML
水杨酸乙酯 ≥99.0% (by GC)
水杨酸乙酯
分子式: C₉H₁₀O₃
沸点: 231°C
溶点: 1 °C
密度: 1,13 g/cm³ (20 °C)
闪点: 120°C
存储条件: Ambient
MDL编号: MFCD00002215
CAS号: 118-61-6
EINECS: 204-265-5
Merck Index: 14,03850
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a7a132d25f8e030c5ff85b9ca0c111d44dd0574&vid=1a7a132d25f8e030c5ff85b9ca0c111d44dd0574&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR