丁酸丁酯 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Butyric acid butyl ester
B0757-25ML
B0757-500ML
TCIAB0757-25MLEA
0
CNY
TCIAB0757-25ML
TCIAB0757-500ML
丁酸丁酯 ≥99.0% (by GC)
丁酸丁酯
分子式:
C₈H₁₆O₂ 分子量: 144.21 g/mol 沸点: 164°C 溶点: -91.5 闪点: 56°C 存储条件: Ambient |
MDL编号:
MFCD00009450 CAS号: 109-21-7 EINECS: 203-656-8 UN号: 3272 ADR: 3 III Merck Index: 14,01556 |
|
产品技术规格
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...