油酰氯 ≥80.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Oleic acid chloride
O0053-25G
O0053-250G
TCIAO0053-25GEA
0
CNY
TCIAO0053-25G
TCIAO0053-250G
油酰氯 ≥80.0% (by GC)
油酰氯
分子式:
C₁₈H₃₃ClO 分子量: 300.91 g/mol 沸点: 378 溶点: -46 闪点: 171°C 存储条件: Refrigerator |
MDL编号:
MFCD00134332 CAS号: 112-77-6 EINECS: 204-005-0 UN号: 3265 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...