苯甲酰氯 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
Benzoic acid chloride
B0105-25ML
B0105-500ML
TCIAB0105-25MLEA
0
CNY
TCIAB0105-25ML
TCIAB0105-500ML
苯甲酰氯 ≥98.0% (by GC, titration analysis)
苯甲酰氯
分子式:
C₇H₅ClO 分子量: 140.57 g/mol 沸点: 196°C 溶点: -0.6 闪点: 72°C 存储条件: Ambient |
MDL编号:
MFCD00000653 CAS号: 98-88-4 EINECS: 202-710-8 UN号: 1736 ADR: 8 II Merck Index: 14,01112 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...