脱氢乙酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-乙酰基-6-甲基-2,4-吡喃二酮

D0039-25G D0039-500G
TCIAD0039-25GEA 0 CNY
TCIAD0039-25G TCIAD0039-500G
脱氢乙酸 ≥98.0% (by GC, titration analysis)
脱氢乙酸
分子式: C₈H₈O₄
分子量: 168.15 g/mol
沸点: 333
溶点: 108
闪点: 157
存储条件: Ambient
MDL编号: MFCD00066709
CAS号: 520-45-6
EINECS: 208-293-9
Merck Index: 14,02865
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a4c6456e7d34c97ae496cdb158044b1d9004d7&vid=46a4c6456e7d34c97ae496cdb158044b1d9004d7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR