茚三酮喷雾显色剂 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 茚三酮

I0015-25G I0015-500G
TCIAI0015-25GEA 0 CNY
TCIAI0015-25G TCIAI0015-500G
茚三酮喷雾显色剂 ≥98.0% (by titrimetric analysis)
茚三酮喷雾显色剂
分子式: C₉H₆O₄
沸点: 351 °C (1013 hPa)
溶点: 250…258 °C
密度: 0,862 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003791
CAS号: 485-47-2
EINECS: 207-618-1
Merck Index: 14,06554
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f6a16695e76f490f3543afea5449d863358936d&vid=6f6a16695e76f490f3543afea5449d863358936d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Oxime Formation) min. 98.0 %
Solubility in Water almost transparency
Ignition residue (Sulfate) max. 0.2 %
suitability for amino acid analysis to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR