5-硝基靛红 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-Nitro-1H-indole-2,3-dione, 5-Nitroindoline-2,3-dione
N0572-25G
N0572-5G
TCIAN0572-25GEA
0
CNY
TCIAN0572-25G
TCIAN0572-5G
5-硝基靛红 ≥98.0% (by HPLC, titration analysis)
5-硝基靛红
分子式:
C₈H₄N₂O₄ 分子量: 192.13 g/mol |
MDL编号:
MFCD00005720 CAS号: 611-09-6 EINECS: 210-252-5 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.000 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in N,N-DMF | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...