9,10-菲醌 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 9,10-菲二酮

P0080-25G P0080-5G
TCIAP0080-25GEA 0 CNY
TCIAP0080-25G TCIAP0080-5G
9,10-菲醌 ≥99.0% (by GC)
9,10-菲醌
分子式: C₁₄H₈O₂
分子量: 208.22 g/mol
沸点: 360
存储条件: Ambient
MDL编号: MFCD00001163
CAS号: 84-11-7
EINECS: 201-515-5
UN号: 3077
ADR: 9 III
Merck Index: 14,07213
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e04f310e4f34d82b13b6dc89d031af6034bc288f&vid=e04f310e4f34d82b13b6dc89d031af6034bc288f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 209.0 to 213.0 deg-C
Solubility in hot DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR