环庚三烯酚酮 ≥98.0% (by GC, titration analysis)
供应商: TCI
Synonyms:
2-羟基-2,4,6-环庚三烯-1-酮
T0606-1G
T0606-25G
T0606-5G
TCIAT0606-1GEA
0
CNY
TCIAT0606-1G
TCIAT0606-25G
TCIAT0606-5G
环庚三烯酚酮 ≥98.0% (by GC, titration analysis)
环庚三烯酚酮
分子式:
C₇H₆O₂ 沸点: 66 - 67 °C (0,1 -rr) 溶点: 48…50 °C 存储条件: Refrigerator |
MDL编号:
MFCD00004158 CAS号: 533-75-5 EINECS: 208-577-2 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...