2-Hydroxychalcone ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3-(2-Hydroxyphenyl)-1-phenylprop-2-en-1-one, 2-Hydroxybenzylidenacetophenone
H0234-25G
H0234-5G
TCIAH0234-25GEA
0
CNY
TCIAH0234-25G
TCIAH0234-5G
2-Hydroxychalcone ≥98.0% (by GC, titration analysis)
2-Hydroxychalcone
分子式:
C₁₅H₁₂O₂ 分子量: 224.26 g/mol |
MDL编号:
MFCD00016449 CAS号: 644-78-0 EINECS: 211-422-1 |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...