2-氮己环酮 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-哌啶酮
P0455-25G
P0455-500G
TCIAP0455-25GEA
0
CNY
TCIAP0455-25G
TCIAP0455-500G
2-氮己环酮 ≥98.0% (by GC)
2-氮己环酮
分子式:
C₅H₉NO 分子量: 99.13 g/mol 沸点: 256 存储条件: Ambient |
MDL编号:
MFCD00006037 CAS号: 675-20-7 EINECS: 211-622-9 |
|
产品技术规格
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...