2'-甲基苯乙酮 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-(2-甲基苯基)乙酮, 邻甲基苯乙酮, Methyl o-tolyl ketone, 1-(2-甲苯基)乙酮, 2-乙酰甲苯

M0172-25ML M0172-500ML
TCIAM0172-25MLEA 0 CNY
TCIAM0172-25ML TCIAM0172-500ML
2'-甲基苯乙酮 ≥98.0% (by GC)
2'-甲基苯乙酮
分子式: C₉H₁₀O
分子量: 134.18 g/mol
沸点: 216°C
闪点: 76°C
存储条件: Ambient
MDL编号: MFCD00008734
CAS号: 577-16-2
EINECS: 209-408-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426cb921b05ebd3312192c01be4097714ba59c3f&vid=426cb921b05ebd3312192c01be4097714ba59c3f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR