2,4-二氯-5-硝基嘧啶 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
D3721-25G
D3721-5G
TCIAD3721-25GEA
0
CNY
TCIAD3721-25G
TCIAD3721-5G
2,4-二氯-5-硝基嘧啶 ≥98.0% (by GC)
2,4-二氯-5-硝基嘧啶
分子式:
C₄HCl₂N₃O₂ 分子量: 193.98 g/mol 沸点: 139°C 溶点: 29.3 密度: 1.74 存储条件: Refrigerator |
MDL编号:
MFCD00127867 CAS号: 49845-33-2 |
|
产品技术规格
Appearance | Light yellow to Yellow to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...