二氟氯乙酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Difluorochloroacetic acid

C0990-25G C0990-500G
TCIAC0990-25GEA 0 CNY
TCIAC0990-25G TCIAC0990-500G
二氟氯乙酸 ≥98.0% (by titrimetric analysis)
二氟氯乙酸
分子式: C₂HClF₂O₂
分子量: 130.48 g/mol
沸点: 126°C
存储条件: Ambient
MDL编号: MFCD00004176
CAS号: 76-04-0
EINECS: 200-928-8
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806e8a981c6efc5ac8cb6d2ce4cc5920e9a30cc7&vid=806e8a981c6efc5ac8cb6d2ce4cc5920e9a30cc7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Brown powder to lump
Purity (Neutralization titration) min. 98.0 %
Freezing point 20.0 to 24.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR