N-氯代丁二酰亚胺 ≥98.0% (by titrimetric analysis) for synthesis
供应商: TCI
|
Danger
|
Synonyms:
1-Chloropyrrolidine-2,5-dione
C0291-25G
C0291-500G
TCIAC0291-25GEA
0
CNY
TCIAC0291-25G
TCIAC0291-500G
N-氯代丁二酰亚胺 ≥98.0% (by titrimetric analysis) for synthesis
N-氯代丁二酰亚胺
分子式:
C₄H₄ClNO₂ 分子量: 133.53 g/mol 沸点: 216.5 存储条件: Refrigerator |
MDL编号:
MFCD00005511 CAS号: 128-09-6 EINECS: 204-878-8 UN号: 3261 ADR: 8 II Merck Index: 14,02164 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 146.0 to 151.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...