苄基三乙基氯化铵 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Benzyl triethylammonium chloride

B0444-25G B0444-500G
TCIAB0444-25GEA 0 CNY
TCIAB0444-25G TCIAB0444-500G
苄基三乙基氯化铵 ≥98.0% (by titrimetric analysis)
苄基三乙基氯化铵
分子式: C₁₃H₂₂ClN
分子量: 227.78 g/mol
存储条件: Ambient
MDL编号: MFCD00011824
CAS号: 56-37-1
EINECS: 200-270-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21b56b37fb0edc45c1c0ce83cb3080116f6938a&vid=b21b56b37fb0edc45c1c0ce83cb3080116f6938a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR