3,5,5-三甲基己醛 ≥90.0% (by GC)

供应商: TCI
Warning

Synonyms: 3,5,5-Trimethylhexanaldehyde

T0940-25ML T0940-500ML
TCIAT0940-25MLEA 0 CNY
TCIAT0940-25ML TCIAT0940-500ML
3,5,5-三甲基己醛 ≥90.0% (by GC)
3,5,5-三甲基己醛
分子式: C₉H₁₈O
沸点: 167°C
闪点: 46°C
MDL编号: MFCD00010441
CAS号: 5435-64-3
EINECS: 226-603-0
UN号: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=326f2931cc7abc228c67e22d7f07a7492af86399&vid=326f2931cc7abc228c67e22d7f07a7492af86399&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR