4-溴-2,6-二甲基苯甲醚 ≥96.0% (by GC)

供应商: TCI
Warning

B3016-25G B3016-5G
TCIAB3016-25GEA 0 CNY
TCIAB3016-25G TCIAB3016-5G
4-溴-2,6-二甲基苯甲醚 ≥96.0% (by GC)
4-溴-2,6-二甲基苯甲醚
分子式: C₉H₁₁BrO
分子量: 215.09 g/mol
沸点: 120°C
MDL编号: MFCD00143259
CAS号: 14804-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19b74fcfa2b5332cde9b772994f83e97f01c4026&vid=19b74fcfa2b5332cde9b772994f83e97f01c4026&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR