N-溴琥珀酰亚胺 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: NBS

B0656-100G B0656-25G B0656-500G
TCIAB0656-100GEA 0 CNY
TCIAB0656-100G TCIAB0656-25G TCIAB0656-500G
N-溴琥珀酰亚胺 ≥98.0% (by titrimetric analysis)
N-溴琥珀酰亚胺
分子式: C₄H₄BrNO₂
沸点: 339°C
溶点: 176°C
密度: 2,098 g/cm³ (20 °C)
存储条件: Refrigerator
MDL编号: MFCD00005510
CAS号: 128-08-5
EINECS: 204-877-2
UN号: 3261
ADR: 8,III
Merck Index: 14,01438
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbc6e996a0b8c4daa176ae1f1256667b75c3e15f&vid=bbc6e996a0b8c4daa176ae1f1256667b75c3e15f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR