2-Amino-3-bromo-5-chloropyridine ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-Bromo-5-chloro-2-pyridinamine, 3-Bromo-5-chloro-2-pyridylamine, 3-Bromo-5-chloropyridin-2-amine

A2402-25G A2402-5G
TCIAA2402-25GEA 0 CNY
TCIAA2402-25G TCIAA2402-5G
2-Amino-3-bromo-5-chloropyridine ≥98.0% (by GC)
2-Amino-3-bromo-5-chloropyridine
分子式: C₅H₄BrClN₂
分子量: 207.46 g/mol
MDL编号: MFCD00955627
CAS号: 26163-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c9a324c67736c3abfc7f1294a898bedced02a4d&vid=9c9a324c67736c3abfc7f1294a898bedced02a4d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR