苄基三乙基溴化铵 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

B0443-25G B0443-500G
TCIAB0443-25GEA 0 CNY
TCIAB0443-25G TCIAB0443-500G
苄基三乙基溴化铵 ≥98.0% (by titrimetric analysis)
苄基三乙基溴化铵
分子式: C₁₃H₂₂BrN
溶点: 195 °C
存储条件: Ambient
MDL编号: MFCD00011822
CAS号: 5197-95-5
EINECS: 225-986-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e1e91c85c7dd89d436d8f1972ccd4a12ad83fc&vid=27e1e91c85c7dd89d436d8f1972ccd4a12ad83fc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR