1,4-Di(bromomethyl)-2,5-dimethylbenzene ≥98.0% (通过GC)

供应商: TCI
Danger

B2210-1G B2210-5G
TCIAB2210-1GEA 0 CNY
TCIAB2210-1G TCIAB2210-5G
1,4-Di(bromomethyl)-2,5-dimethylbenzene ≥98.0% (通过GC)
1,4-Di(bromomethyl)-2,5-dimethylbenzene
分子式: C₁₀H₁₂Br₂
MDL编号: MFCD04038409
CAS号: 35168-62-8
EINECS: 252-413-2
UN号: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24ee9ec488ec55dea0cf9e328e1990b608da1d42&vid=24ee9ec488ec55dea0cf9e328e1990b608da1d42&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 159.0 to 163.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR