羟胺-O-磺酸 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Amino hydrogen sulphate
325940250
325941000
ACRO325940250EA
0
CNY
ACRO325940250
ACRO325941000
羟胺-O-磺酸 97%
羟胺-O-磺酸
分子式:
H₃NO₄S 分子量: 113.09 g/mol 溶点: 210 °C 存储条件: Refrigerator |
MDL编号:
MFCD00011604 CAS号: 2950-43-8 EINECS: 220-971-6 UN号: 3260 ADR: 8 II |
产品技术规格
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...