苯甲酰甲酸乙酯 95%
供应商: Thermo Fisher Scientific
Synonyms:
α-羰基苯乙酸乙酯
410350050
410350250
ACRO410350050EA
0
CNY
ACRO410350050
ACRO410350250
苯甲酰甲酸乙酯 95%
苯甲酰甲酸乙酯
分子式:
C₁₀H₁₀O₃ 分子量: 178.19 g/mol 沸点: 138…139 °C (18 mmHg) 闪点: 113 °C 存储条件: Ambient |
MDL编号:
MFCD00009120 CAS号: 1603-79-8 EINECS: 216-504-0 |
产品技术规格
Appearance | Clear colorless to green Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5150 to 1.5170 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...