亚氨基二乙酸 98+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,2'-Azanediyldiacetic acid
204970250
204975000
ACRO204970250EA
0
CNY
ACRO204970250
ACRO204975000
亚氨基二乙酸 98+%
亚氨基二乙酸
分子式:
C₄H₇NO₄ 溶点: 243 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00004280 CAS号: 142-73-4 EINECS: 205-555-4 |
产品技术规格
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...