N-乙氧甲酰邻苯二甲酰亚胺 99+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Ethyl phthalimide-N-carboxylate, N-乙氧羰基邻苯二甲酰亚胺, Ethyl 1,3-dioxo-2-isoindolinecarboxylate
108310500
108310100
ACRO108310500EA
0
CNY
ACRO108310500
ACRO108310100
N-乙氧甲酰邻苯二甲酰亚胺 99+%
N-乙氧甲酰邻苯二甲酰亚胺
分子式:
C₁₁H₉NO₄ 分子量: 219.20 g/mol 溶点: 82…84 °C 存储条件: Ambient |
MDL编号:
MFCD00005893 CAS号: 22509-74-6 EINECS: 245-048-5 |
产品技术规格
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 82 to 84 °C |
Water | ≤0.5 % (K.F.) |
Titration | ≥99.0 % (Saponification) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...