3-呋喃硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 呋喃-3-硼酸

F0438-1G F0438-5G
TCIAF0438-1GEA 0 CNY
TCIAF0438-1G TCIAF0438-5G
3-呋喃硼酸 (contains varying amounts of Anhydride)
3-呋喃硼酸
分子式: C₄H₅BO₃
MDL编号: MFCD01319007
CAS号: 55552-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443693a2148d1c8fb59c32f90af0793e394a2096&vid=443693a2148d1c8fb59c32f90af0793e394a2096&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 to 119.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR