(S)-(+)-2-丁醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(S)-(+)-仲丁基醇
B0925-1ML
B0925-5ML
TCIAB0925-1MLEA
0
CNY
TCIAB0925-1ML
TCIAB0925-5ML
(S)-(+)-2-丁醇 ≥98.0% (by GC)
(S)-(+)-2-丁醇
分子式:
C₄H₁₀O 分子量: 74.12 g/mol 沸点: 100°C 溶点: -114 闪点: 24°C 存储条件: Ambient |
MDL编号:
MFCD00064281 CAS号: 4221-99-2 EINECS: 224-168-1 UN号: 1120 ADR: 3 III Merck Index: 14,01541 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +11.0 to +14.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...