1-萘基异氰酸酯 ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: 1-Naphthylisocyanate

I0125-25ML I0125-5ML
TCIAI0125-25MLEA 0 CNY
TCIAI0125-25ML TCIAI0125-5ML
1-萘基异氰酸酯 ≥99.0% (by GC)
1-萘基异氰酸酯
分子式: C₁₁H₇NO
分子量: 169.18 g/mol
沸点: 269°C
溶点: 36
密度: 1.18
闪点: 135°C
存储条件: Refrigerator
MDL编号: MFCD00003881
CAS号: 86-84-0
EINECS: 201-703-7
UN号: 2206
ADR: 6.1 III
Merck Index: 14,06410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57d9eced9400a1c713ae805594f17cb6c06abe84&vid=57d9eced9400a1c713ae805594f17cb6c06abe84&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR