Adapters, tubing, KA, KECK™, DURAN®
供应商: DWK LIFE SCIENCES
286345105 286345002 286345208 286345302
SCOT286345105EA 0 CNY
SCOT286345105 SCOT286345002 SCOT286345208 SCOT286345302
Adapters, tubing, KA, KECK™, DURAN®
接头
KECK™ adapter for glass thread KA with screw threaded cap.
  • Connect or remove hose by twisting 1/4 turn
  • Available for sizes GL 14, RD 14 and SQ 13
It protects the thread from damage and guarantee fast, safe and leakproof (upto 30 bar pressure and 0,66 bar vacuum) connections.
Delivery information: The adapters are available as a complete set which comes either with 4,5 / 9,0 mm Ø tubing part.