2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid), Ultrapure
供应商: VWR Chemicals
|
Warning
|
Synonyms:
AzBTS-(NH4)2, Diammonium 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulfonate)
VWRC0400-100GEA
65178
CNY
VWRC0400-100G
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid), Ultrapure
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid)
Substrate for the detection of horseradish peroxidase commonly used in ELISA. Read at 450 nm.
Soluble.reaction condition: 1 mM in 50 mM sodium citrate, 150 mM sodium phosphate and 0.01% H₂O₂. Stop with 1.5 mM NaN₃ in 0.1 M citric acid.
分子式:
C₁₈H₂₄N₆O₆S₄ 分子量: 548.68 g/mol 溶点: >300°C 存储条件: Refrigerator |
MDL编号:
MFCD00010404 CAS号: 30931-67-0 EINECS: 250-396-6 |
产品技术规格
E max @225 nm | >48150 |
E max @342 nm | ≥37400 |
Free ammonia | ≤6.5% |
Loss on drying | ≤2% |
Solubility (5%, Water) | Pass |
Thin layer chromatography | ≥98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...