Hydroxypropyl cellulose, M.W. ∼ 100.000
供应商: Thermo Fisher Scientific
184881000
184882500
ACRO184881000EA
0
CNY
ACRO184881000
ACRO184882500
Hydroxypropyl cellulose, M.W. ∼ 100.000
Hydroxypropyl cellulose
存储条件:
Ambient |
MDL编号:
MFCD00132688 CAS号: 9004-64-2 |
产品技术规格
Appearance | White to cream Crystalline powder or granules |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % (105°C) |
Sulfated ash | ≤0.5 % |
Viscosity | 75 to 150 mPa.s (5 % in water) (Brookfield settings: 30 rpm, spindle no.1) |
pH | 5 to 8.5 (1 % solution) |
Arsenic (As) | ≤3 ppm |
Lead (Pb) | ≤10 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...