Temperature probes, platinum resistance
供应商: COWIE
500.200.1 500.100.1 500.300.1 500.400.1 500.500.1 500.600.1 500.700.1 500.900.1 500.307.1 500.208.1 500.206.1 500.106.1
COWI500.200.1EA 0 CNY
COWI500.200.1 COWI500.100.1 COWI500.300.1 COWI500.400.1 COWI500.500.1 COWI500.600.1 COWI500.700.1 COWI500.900.1 COWI500.307.1 COWI500.208.1 COWI500.206.1 COWI500.106.1
Temperature probes, platinum resistance
温度探头
Platinum resistance temperature probes made of stainless steel, encapsulated in PTFE, with push-on ferrule.
  • Inert and non contaminating
  • Pressure resistent
  • Features 1 m long PFA insulated cable
Response: 50%, 6 to 14 seconds
Delivery information: Supplied without plug.