6-氨基-1,4-苯并二氧杂环 99%

供应商: Thermo Fisher Scientific
Warning

185780500 185780050
ACRO185780500EA 0 CNY
ACRO185780500 ACRO185780050
6-氨基-1,4-苯并二氧杂环 99%
分子式: C₈H₉NO₂
密度: 1,23 g/cm³
闪点: 120 °C
存储条件: Ambient
MDL编号: MFCD00006824
CAS号: 22013-33-8
EINECS: 244-718-4
UN号: 2811
ADR: 6.1,III

现在订购

产品技术规格

Appearance Yellow to dark brown Solid or liquid
Infrared spectrum Conforms
Refractive index 1.5970 to 1.6010 (20°C, 589 nm)
Titration with HClO4 94.0 to 106.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR