N,N,N′,N′′,N′′-五甲基二乙烯三胺 98+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
PMDETA, PMDTA
416950250
416951000
416955000
ACRO416950250EA
0
CNY
ACRO416950250
ACRO416951000
ACRO416955000
N,N,N′,N′′,N′′-五甲基二乙烯三胺 98+%
N,N,N′,N′′,N′′-五甲基二乙烯三胺
分子式:
C₉H₂₃N₃ 分子量: 173.30 g/mol 溶点: 18 °C 闪点: 72 °C 存储条件: Ambient |
MDL编号:
MFCD00014876 CAS号: 3030-47-5 EINECS: 221-201-1 UN号: 2922 ADR: 8 II |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4415 to 1.4435 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...