Safety shields, SP Bel-Art
供应商: Bel-Art Products, a Part of SP
H249640000 H249620000
BELAH249640000EA 0 CNY
BELAH249640000 BELAH249620000
Safety shields, SP Bel-Art
防护罩 安全防护罩
The stable transparent safety shield PC (thickness: 4,7 mm) is constructed of clear PC barrier and heavy steel base is coated with blue epoxy coating.
  • Three-sided construction provides front and side protection against flying debris
  • Plated steel bolts, nuts and washers ensure long life and rust-resistance
Certifications: ANSI Z87.1, 9.11, 9.12