4,4'-二羟基二苯硫醚 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
TDP, 4,4′-硫代二苯酚
D1356-25G
D1356-500G
TCIAD1356-25GEA
0
CNY
TCIAD1356-25G
TCIAD1356-500G
4,4'-二羟基二苯硫醚 ≥98.0% (by GC)
4,4'-二羟基二苯硫醚
分子式:
C₁₂H₁₀O₂S 分子量: 218.28 g/mol |
MDL编号:
MFCD00002349 CAS号: 2664-63-3 EINECS: 220-197-9 |
|
产品技术规格
Appearance | White to Light yellow powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...