顺式乌头酸, Sigma-Aldrich®

供应商: Merck

Synonyms: 顺式-1,2,3-丙烯三羧酸

M-Clarity™ quality level = MQ200

A3412-1G A3412-5G
SIALA3412-1GEA 0 CNY
SIALA3412-1G SIALA3412-5G
顺式乌头酸, Sigma-Aldrich®
顺式乌头酸
分子式: C₆H₆O₆
分子量: 174.11 g/mol
MDL编号: MFCD00063184
CAS号: 585-84-2
EINECS: 209-564-4

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR