(3-Chlorophenyl)acetone 94%
供应商: Thermo Fisher Scientific
Synonyms:
1-(3-Chlorophenyl)propan-2-one
350690010
350690050
ACRO350690010EA
0
CNY
ACRO350690010
ACRO350690050
(3-Chlorophenyl)acetone 94%
(3-Chlorophenyl)acetone
分子式:
C₉H₉ClO 存储条件: Ambient |
MDL编号:
MFCD00082872 CAS号: 14123-60-5 |
产品技术规格
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5320 to 1.5360 (20°C, 589 nm) |
GC | ≥92 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...