甲基丙二酸二乙酯 99%
供应商: Thermo Fisher Scientific
Synonyms:
Methylmalonic acid diethyl ester
155141000
155145000
ACRO155141000EA
0
CNY
ACRO155141000
ACRO155145000
甲基丙二酸二乙酯 99%
甲基丙二酸二乙酯
分子式:
C₈H₁₄O₄ 密度: 1,01 g/cm³ 闪点: 76 °C 存储条件: Ambient |
MDL编号:
MFCD00009162 CAS号: 609-08-5 EINECS: 210-175-7 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4120 to 1.4140 (20°C, 589 nm) |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...