Sodium hydroxide 10 mol/l (10 N) in aqueous solution 10 mol/l (10 N) in aqueous solution, Reagent Grade
供应商: VWR Chemicals
Danger
Synonyms: 苛性钠
VWRCE584-100MLEA 686 CNY
VWRCE584-100ML VWRCE584-500ML
Sodium hydroxide 10 mol/l (10 N) in aqueous solution 10 mol/l (10 N) in aqueous solution, Reagent Grade
氢氧化钠
分子式: NaOH
分子量: 40 g/mol
沸点: 1390
溶点: 323
存储条件: Ambient
MDL编号: MFCD00003548
CAS号: 1310-73-2
EINECS: 215-185-5
UN号: 1824
ADR: 8 II
Merck Index: 14,08627
现在订购
产品技术规格
Acid Titration 9900 mmol/l – 10100 mmol/l