Sodium cholate, Reagent Grade

供应商: VWR Chemicals

VWRC0152-100GEA 1270 CNY
VWRC0152-100G VWRC0152-1KG
Sodium cholate, Reagent Grade
Sodium cholate
分子式: C₂₄H₃₉NaO₅
溶点: 198 °C
存储条件: Ambient
MDL编号: MFCD00150749
CAS号: 361-09-1
EINECS: 206-643-5

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR