VWR®, Precut Pierceable Vinyl Films for Robotics
供应商: VWR Collection
VWRI391-1286EA 3435 CNY
VWRI391-1286 VWRI391-1287
VWR®, Precut Pierceable Vinyl Films for Robotics
密封件 微孔板封板膜和铝箔
该款106,7 µm厚的丁基膜附有20 µm粘合剂层,用于临时保护96孔板免受污染和免于蒸发.
  • 能够短期保护样品并减少蒸发
  • 预裁切内弯瓣,避免污染探头或吸头
  • 取样后弹性瓣闭合,继续保护样品
  • 建议使用温度范围−40 到+90 °C
每个孔上的预切图案将膜分隔成四瓣,当受到来自机械探头或移液器吸头压力时可轻松内弯,无需钻孔或受到胶黏剂污染便可获取样品。完成取样后,弹性瓣可以恢复原来位置,继续保护样品。为了取样后长期保护样品,应覆上连续膜作为第二层。适用于密封所有标准96孔板.
每个膜的长×深:145,5×79,4 mm