Culture media flasks, DURAN®
供应商: DWK LIFE SCIENCES
214314401 214315406
SCOT214314401EA 0 CNY
SCOT214314401 SCOT214315406
Culture media flasks, DURAN®
培养基瓶
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With straight neck for glass caps. Matching glass caps are optionally available. Marked with volume and equipped with a white labelling field.