4-(4-氯苯基)-4-羟基哌啶 99+%, crystalline

供应商: Thermo Fisher Scientific
Warning

Synonyms: 4-(4-氯苯基)-4-羟基哌啶, 4-(4-Chlorophenyl)-4-hydroxypiperidine, 4-(4-氯苯基)-4-哌啶醇

296840010 296840050
ACRO296840010EA 0 CNY
ACRO296840010 ACRO296840050
4-(4-氯苯基)-4-羟基哌啶 99+%, crystalline
4-(4-氯苯基)-4-羟基哌啶
分子式: C₁₁H₁₄ClNO
分子量: 211.69 g/mol
溶点: 139…141 °C
存储条件: Ambient
MDL编号: MFCD00006001
CAS号: 39512-49-7
EINECS: 254-479-8

现在订购

产品技术规格

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 2 h, 760 mmHg)
Sulfated ash ≤0.2 %
TLC ≤0.5 % (Each individual impurity)
UV at 440 nm A: ≤0.05
Titration with HClO4 98.0 to 102.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR