叔丁醇钠 99%, pure

供应商: Thermo Fisher Scientific
Danger

Synonyms: Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate

252550050 252551000
ACRO252550050EA 0 CNY
ACRO252550050 ACRO252551000
叔丁醇钠 99%, pure
叔丁醇钠
分子式: C₄H₉NaO
溶点: 180 °C
闪点: 12 °C
存储条件: Ambient
MDL编号: MFCD00040575
CAS号: 865-48-5
EINECS: 212-741-9
UN号: 3206
ADR: 4.2,II

现在订购

产品技术规格

Appearance White to light brown Powder
Titration with HCl 98.5 to 101.5 %
NaOH + Na2CO3 (as NaOH) ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR