3-甲基-1,3-丁二醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
3-Methylbutane-1,3-diol
M0818-25ML
M0818-500ML
TCIAM0818-25MLEA
0
CNY
TCIAM0818-25ML
TCIAM0818-500ML
3-甲基-1,3-丁二醇 ≥98.0% (by GC)
3-甲基-1,3-丁二醇
分子式:
C₅H₁₂O₂ 沸点: 203°C 闪点: 116°C |
MDL编号:
MFCD00059655 CAS号: 2568-33-4 EINECS: 459-270-7 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...