dNTP solutions, illustra™
供应商: Cytiva
28-4065-42
28-4065-41
28-4065-32
28-4065-22
28-4065-02
28-4065-01
28-4065-12
GEHE28-4065-42EA
0
CNY
GEHE28-4065-42
GEHE28-4065-41
GEHE28-4065-32
GEHE28-4065-22
GEHE28-4065-02
GEHE28-4065-01
GEHE28-4065-12
dNTP solutions, illustra™
核酸试剂
核苷酸
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
规格
Environmentally Preferable | |
Application |
Learn more